siRNA Details
| Gene Symbol: Rasip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028544.1 | NP_082820.1 | 8 | 2142 | ||
| GenBank mRNA | AK011557.2 | - | 2 | 225 | ||
| AK033276.1 | - | 6 | 1430 | |||
| AK156602.1 | - | 8 | 2154 | |||
| BC028444.1 | - | 214 | ||||
| BC072584.1 | - | 5 | 1096 | |||
| BC127059.1 | - | 6 | 1068 | 40109852 | ||
| BC150766.1 | - | 8 | 2142 | 9087677 | ||

Rasip1
Genomic Map