siRNA Details
| Gene Symbol: Nkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027280.3 | NP_081556.3 | 3 | 263 | ||
| GenBank mRNA | AF343352.1 | - | 3 | 100 | ||
| AF358134.1 | - | 3 | 179 | |||
| AK076164.1 | - | 3 | 254 | |||
| AK082367.1 | - | 3 | 251 | |||
| AK166564.1 | - | 3 | 266 | |||
| BC034838.1 | - | 3 | 237 | 1429943 | ||

Nkd1
Genomic Map