siRNA Details
| Gene Symbol: Clic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027085.3 | NP_081361.1 | 4, 5 | 416 | ||
| GenBank mRNA | AK009020.1 | - | 4, 5 | 418 | ||
| BC060967.1 | - | 4, 5 | 421 | 30297308 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clic3| Gene Symbol: Clic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027085.3 | NP_081361.1 | 4, 5 | 416 | ||
| GenBank mRNA | AK009020.1 | - | 4, 5 | 418 | ||
| BC060967.1 | - | 4, 5 | 421 | 30297308 | ||