siRNA Details
| Gene Symbol: Ubl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122873.1 | NP_001116345.1 | 6 | 594 | ||
| NM_027086.3 | NP_081362.2 | 6 | 542 | |||
| GenBank mRNA | AK009025.1 | - | 6 | 543 | ||
| AK154496.1 | - | 6 | 552 | |||
| AK179100.1 | - | 2 | 106 | |||
| AK182178.1 | - | 2 | 106 | |||
| AK207226.1 | - | 2 | 106 | |||
| AK212304.1 | - | 2 | 106 | |||
| BC016456.1 | - | 5 | 528 | 4912715 | ||
| BC094047.1 | - | 5 | 531 | 5253101 | ||
| CX203425.1 | - | 6 | 572 | |||

Ubl7
Genomic Map