siRNA Details
| Gene Symbol: Smyd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027188.3 | NP_081464.1 | 3, 4 | 380 | ||
| GenBank mRNA | AK010447.1 | - | 3, 4 | 414 | ||
| AK043988.1 | - | 3, 4 | 390 | |||
| AK087728.1 | - | 3, 4 | 392 | |||
| AK163057.1 | - | 3, 4 | 397 | |||
| AK167875.1 | - | 3, 4 | 395 | |||
| BC019663.1 | - | 3, 4 | 382 | |||
| BC038482.1 | - | 3, 4 | 261 | |||
| BC052431.1 | - | 3, 4 | 380 | 6837012 | ||
| BC061485.1 | - | 3, 4 | 363 | 5370924 | ||

Smyd3
Genomic Map