siRNA Details
| Gene Symbol: Nup37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027191.2 | NP_081467.2 | 2, 3 | 307 | ||
| NM_028334.4 | NP_082610.1 | 2, 3 | 325 | |||
| GenBank mRNA | AK010370.1 | - | 2, 3 | 251 | ||
| AK012890.1 | - | 2, 3 | 321 | |||
| AK040463.1 | - | 2, 3 | 308 | |||
| AK165537.1 | - | 2, 3 | 271 | |||
| BC011102.1 | - | 2, 3 | 291 | 3982076 | ||
| BC032180.1 | - | 1, 2 | 235 | 5151847 | ||
| CJ072834.1 | - | 2, 3 | 327 | |||

Nup37
Genomic Map