siRNA Details
| Gene Symbol: Clec4b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190310.1 | NP_001177239.1 | 6 | 656 | ||
| NM_027218.3 | NP_081494.1 | 5 | 557 | |||
| GenBank mRNA | AK007794.1 | - | 5 | 514 | ||
| AY230259.1 | - | 6 | 620 | |||
| AY230260.1 | - | 5 | 521 | |||
| AY365134.1 | - | 6 | 552 | |||

Clec4b1
Genomic Map