siRNA Details
Gene Symbol: 1810065E05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027239.2 | NP_081515.2 | 4 | 322 | ||
GenBank mRNA | AK007956.1 | - | 4 | 358 | ||
BC026941.1 | - | 4 | 294 | 4024443 | ||
BY092570.1 | - | 4 | 319 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
