siRNA Details
| Gene Symbol: 2010107G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027251.3 | NP_081527.1 | 4 | 645 | ||
| GenBank mRNA | AK008338.1 | - | 3 | 565 | ||
| AK035688.1 | - | 4 | 646 | |||
| BC024943.1 | - | 4 | 960 | 3989515 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2010107G23Rik| Gene Symbol: 2010107G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027251.3 | NP_081527.1 | 4 | 645 | ||
| GenBank mRNA | AK008338.1 | - | 3 | 565 | ||
| AK035688.1 | - | 4 | 646 | |||
| BC024943.1 | - | 4 | 960 | 3989515 | ||