siRNA Details
| Gene Symbol: 2810025M15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012812.1 | - | 3 | 666 | ||
| BC055845.1 | - | 3 | 1090 | 3585495 | ||
| BC055878.1 | - | 3 | 676 | 4222031 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810025M15Rik| Gene Symbol: 2810025M15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012812.1 | - | 3 | 666 | ||
| BC055845.1 | - | 3 | 1090 | 3585495 | ||
| BC055878.1 | - | 3 | 676 | 4222031 | ||