siRNA Details
| Gene Symbol: Manba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027288.3 | NP_081564.3 | 4 | 597 | ||
| GenBank mRNA | AF306557.1 | - | 4 | 601 | ||
| AK036505.1 | - | 4 | 595 | |||
| AK047057.1 | - | 4 | 590 | |||
| AK142165.1 | - | 4 | 562 | |||
| AK154172.1 | - | 4 | 578 | |||
| AK166022.1 | - | 4 | 579 | |||
| AK166340.1 | - | 4 | 595 | |||
| AK170290.1 | - | 4 | 594 | |||
| AK171416.1 | - | 3 | 1306 | |||
| BC031409.1 | - | 4 | 540 | 4952710 | ||

Manba
Genomic Map