siRNA Details
| Gene Symbol: Krtap26-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027105.2 | NP_081381.1 | 1 | 710 | ||
| GenBank mRNA | AK009086.1 | - | 1 | 712 | ||
| BC128283.1 | - | 1 | 702 | 40101041 | ||
| BC139410.1 | - | 1 | 705 | 8862432 | ||
| BC139411.1 | - | 1 | 705 | 8862433 | ||

Krtap26-1
Genomic Map