siRNA Details
| Gene Symbol: Ddx51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027156.3 | NP_081432.2 | 15 | 2471 | ||
| GenBank mRNA | AK010015.1 | - | 6 | 1075 | ||
| AK141361.1 | - | 15 | 2452 | |||
| AK152599.1 | - | 15 | 2439 | |||
| BC023700.1 | - | 12 | 1734 | |||
| BC060646.1 | - | 15 | 2448 | 6853826 | ||
| BC069876.1 | - | 8 | 1299 | |||

Ddx51
Genomic Map