siRNA Details
| Gene Symbol: Mzb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027222.3 | NP_081498.2 | 4 | 873 | ||
| GenBank mRNA | AA866731.1 | - | 2 | 497 | ||
| AK008016.1 | - | 4 | 874 | |||
| AK088094.1 | - | 4 | 893 | |||
| BC030674.1 | - | 3 | 655 | |||
| BC064044.1 | - | 4 | 860 | 875322 | ||

Mzb1
Genomic Map