siRNA Details
Gene Symbol: 1810065E05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027239.2 | NP_081515.2 | 6 | 860 | ||
GenBank mRNA | AK007956.1 | - | 6 | 896 | ||
BC026941.1 | - | 6 | 832 | 4024443 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1810065E05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027239.2 | NP_081515.2 | 6 | 860 | ||
GenBank mRNA | AK007956.1 | - | 6 | 896 | ||
BC026941.1 | - | 6 | 832 | 4024443 |