siRNA Details
| Gene Symbol: Nkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163660.1 | NP_001157132.1 | 7 | 725 | ||
| NM_027280.3 | NP_081556.3 | 9 | 935 | |||
| GenBank mRNA | AF343352.1 | - | 9 | 772 | ||
| AF358134.1 | - | 9 | 851 | |||
| AK076164.1 | - | 9 | 926 | |||
| AK082367.1 | - | 9 | 924 | |||
| AK166564.1 | - | 9 | 938 | |||
| AK189531.1 | - | 1 | 94 | |||
| AK190160.1 | - | 2 | 125 | |||
| BC034838.1 | - | 9 | 909 | 1429943 | ||
| DV662736.1 | - | 7 | 705 | |||

Nkd1
Genomic Map