siRNA Details
| Gene Symbol: Isl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027397.3 | NP_081673.2 | 4 | 875 | ||
| GenBank mRNA | AK013964.1 | - | 4 | 818 | ||
| BC120880.2 | - | 3 | 555 | |||
| BC148307.2 | - | 3 | 555 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Isl2| Gene Symbol: Isl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027397.3 | NP_081673.2 | 4 | 875 | ||
| GenBank mRNA | AK013964.1 | - | 4 | 818 | ||
| BC120880.2 | - | 3 | 555 | |||
| BC148307.2 | - | 3 | 555 | |||