siRNA Details
| Gene Symbol: Epc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007935.2 | NP_031961.1 | 2 | 383 | ||
| NM_027497.3 | NP_081773.1 | 2 | 534 | |||
| GenBank mRNA | AF079765.1 | - | 2 | 362 | ||
| AK040254.1 | - | 2 | 534 | |||
| AK148194.1 | - | 2 | 352 | |||
| AK160362.1 | - | 2 | 497 | |||
| BC138622.1 | - | 2 | 362 | 8861642 | ||
| BC138623.1 | - | 2 | 362 | 8861643 | ||

Epc1
Genomic Map