siRNA Details
| Gene Symbol: Tspan2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243132.1 | NP_001230061.1 | 2 | 158 | ||
| NM_027533.3 | NP_081809.2 | 2 | 353 | |||
| GenBank mRNA | AK018178.1 | - | 2 | 156 | ||
| AK020982.1 | - | 2 | 158 | |||
| AK031683.1 | - | 1 | 62 | |||
| AK032520.1 | - | 2 | 179 | |||
| AK049888.1 | - | 2 | 155 | |||
| AK079560.1 | - | 2 | 157 | |||
| BB642061.1 | - | 2 | 354 | |||
| BC007185.1 | - | 2 | 594 | 3591472 | ||

Tspan2
Genomic Map