siRNA Details
| Gene Symbol: Prdm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027547.2 | NP_081823.2 | 1, 2 | 240 | ||
| GenBank mRNA | AK014501.1 | - | 1, 2 | 164 | ||
| AK018310.1 | - | 1, 2 | 178 | |||
| AK200908.1 | - | 1, 2 | 87 | |||
| BC089598.1 | - | 1, 2 | 180 | |||
| BC138901.1 | - | 1, 2 | 214 | 8861923 | ||
| BC138902.1 | - | 1, 2 | 214 | 8861924 | ||
| BY014014.1 | - | 1, 2 | 241 | |||

Prdm5
Genomic Map