siRNA Details
| Gene Symbol: Mllt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286158.1 | NP_001273087.1 | 2, 3 | 197 | ||
| NM_027326.3 | NP_081602.3 | 6, 7 | 1524 | |||
| NM_029931.3 | NP_084207.2 | 2, 3 | 197 | |||
| GenBank mRNA | AF333960.1 | - | 5, 6 | 1185 | ||
| AK008707.1 | - | 2, 3 | 159 | |||
| AK019458.1 | - | 2, 3 | 189 | |||
| AK136066.1 | - | 2, 3 | 757 | |||
| AK147009.1 | - | 2, 3 | 111 | |||
| BC021420.1 | - | 7, 8 | 1363 | 5012671 | ||
| BU614509.1 | - | 2, 3 | 325 | |||
| BY713607.1 | - | 2, 3 | 185 | |||
| DV051568.1 | - | 207 | ||||

Mllt3
Genomic Map