siRNA Details
| Gene Symbol: Ufsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027356.2 | NP_081632.1 | 1 | 522 | ||
| GenBank mRNA | AK012343.1 | - | 1 | 523 | ||
| BC016577.1 | - | 1 | 531 | 3584484 | ||
| BC087958.1 | - | 1 | 543 | 6751787 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ufsp1| Gene Symbol: Ufsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027356.2 | NP_081632.1 | 1 | 522 | ||
| GenBank mRNA | AK012343.1 | - | 1 | 523 | ||
| BC016577.1 | - | 1 | 531 | 3584484 | ||
| BC087958.1 | - | 1 | 543 | 6751787 | ||