siRNA Details
| Gene Symbol: Ube2q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027315.4 | NP_081591.3 | 13 | 1632 | ||
| GenBank mRNA | AK009324.1 | - | 12 | 1214 | ||
| AK146707.1 | - | 13 | 1445 | |||
| AK211403.1 | - | 1 | 52 | |||
| AY112699.1 | - | 13 | 1624 | |||
| BC030044.1 | - | 10 | 957 | |||
| BC051487.1 | - | 6 | 629 | |||
| BC060966.1 | - | 11 | 1025 | |||
| BC082275.1 | - | 13 | 1455 | 6310111 | ||

Ube2q1
Genomic Map