siRNA Details
| Gene Symbol: Fam162a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027342.1 | NP_081618.1 | 4 | 329 | ||
| GenBank mRNA | AK009946.1 | - | 4 | 329 | ||
| AK182371.1 | - | 1 | 17 | |||
| AK212003.1 | - | 1 | 17 | |||
| BC010826.1 | - | 4 | 338 | 4219531 | ||
| M97159.1 | - | 300 | ||||

Fam162a
Genomic Map