siRNA Details
| Gene Symbol: PEX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000288.3 | NP_000279.1 | 3, 4 | 446 | ||
| XM_005267019.4 | XP_005267076.1 | 3, 4 | 726 | |||
| XM_017010934.1 | XP_016866423.1 | 3, 4 | 790 | |||
| GenBank mRNA | BC006268.1 | - | 3, 4 | 388 | 3948721 | |
| BC031606.1 | - | 3, 4 | 430 | 5176358 | ||
| BP246238.1 | - | 3, 4 | 446 | |||
| CR542054.1 | - | 3, 4 | 344 | |||
| CR542076.1 | - | 3, 4 | 344 | |||
| U76560.1 | - | 3, 4 | 392 | |||
| U88871.1 | - | 3, 4 | 425 | |||

PEX7
Genomic Map