siRNA Details
| Gene Symbol: Steap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027399.3 | NP_081675.2 | 3, 4 | 707 | ||
| GenBank mRNA | AF297098.1 | - | 2, 3 | 637 | ||
| AK010437.1 | - | 3, 4 | 702 | |||
| AK144312.1 | - | 3, 4 | 662 | |||
| BC061023.1 | - | 3, 4 | 692 | 30304473 | ||

Steap1
Genomic Map