siRNA Details
| Gene Symbol: Rufy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027425.3 | NP_081701.2 | 14 | 1509 | ||
| GenBank mRNA | AK080289.1 | - | 1 | 425 | ||
| AK173195.1 | - | 14 | 1470 | |||
| BC137736.1 | - | 14 | 1458 | 8860755 | ||
| BC137737.1 | - | 14 | 1458 | 8860756 | ||

Rufy2
Genomic Map