siRNA Details
| Gene Symbol: PMVK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323011.1 | NP_001309940.1 | 2, 3 | 140 | ||
| NM_001323012.1 | NP_001309941.1 | 2, 3 | 147 | |||
| NM_006556.3 | NP_006547.1 | 2, 3 | 490 | |||
| GenBank mRNA | AK130234.1 | - | 3, 4 | 332 | ||
| AK312803.1 | - | 2, 3 | 449 | |||
| BC006089.1 | - | 2, 3 | 473 | 3604167 | ||
| BC007694.2 | - | 2, 3 | 403 | 3954909 | ||
| BQ061044.1 | - | 2, 3 | 490 | |||
| BT019976.1 | - | 2, 3 | 167 | |||
| CR541998.1 | - | 2, 3 | 167 | |||
| L77213.1 | - | 2, 3 | 202 | |||

PMVK
Genomic Map