siRNA Details
| Gene Symbol: Fam213a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027464.3 | NP_081740.2 | 6 | 1435 | ||
| GenBank mRNA | AK017688.1 | - | 6 | 1436 | ||
| AK156326.1 | - | 6 | 1469 | |||
| BC056635.1 | - | 6 | 1426 | 5064081 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam213a| Gene Symbol: Fam213a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027464.3 | NP_081740.2 | 6 | 1435 | ||
| GenBank mRNA | AK017688.1 | - | 6 | 1436 | ||
| AK156326.1 | - | 6 | 1469 | |||
| BC056635.1 | - | 6 | 1426 | 5064081 | ||