siRNA Details
| Gene Symbol: Ankrd24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027480.3 | NP_081756.4 | 17 | 1791 | ||
| GenBank mRNA | AK019475.1 | - | 4 | 457 | ||
| AK043638.1 | - | 15 | 2873 | |||
| AK173316.1 | - | 15 | 2757 | |||
| BC017633.1 | - | 4 | 462 | |||
| BC048236.1 | - | 16 | 1696 | 6313999 | ||

Ankrd24
Genomic Map