siRNA Details
| Gene Symbol: Zcchc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027494.3 | NP_081770.3 | 5, 6 | 620 | ||
| GenBank mRNA | AK017857.1 | - | 5, 6 | 516 | ||
| AK032349.1 | - | 5, 6 | 597 | |||
| AK033469.1 | - | 5, 6 | 570 | |||
| AK156770.1 | - | 5, 6 | 592 | |||
| AK166990.1 | - | 5, 6 | 527 | |||
| AK167126.1 | - | 5, 6 | 525 | |||
| AK167736.1 | - | 5, 6 | 527 | |||
| BC013555.1 | - | 4, 5 | 272 | 4238634 | ||

Zcchc8
Genomic Map