siRNA Details
| Gene Symbol: Armc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030184.2 | NP_084460.1 | 24 | 2443 | ||
| GenBank mRNA | AK019600.1 | - | 24 | 2432 | ||
| AK036083.1 | - | 24 | 2411 | |||
| AK046693.1 | - | 24 | 2429 | |||
| BC043101.1 | - | 24 | 2395 | 6406321 | ||

Armc9
Genomic Map