siRNA Details
| Gene Symbol: Prdm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027547.2 | NP_081823.2 | 7, 8 | 995 | ||
| GenBank mRNA | AK014501.1 | - | 7, 8 | 919 | ||
| BC138901.1 | - | 7, 8 | 969 | 8861923 | ||
| BC138902.1 | - | 7, 8 | 969 | 8861924 | ||

Prdm5
Genomic Map