siRNA Details
| Gene Symbol: Dmrtc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027591.3 | NP_081867.1 | 2 | 172 | ||
| GenBank mRNA | AK014934.1 | - | 2 | 173 | ||
| BC147175.1 | - | 2 | 144 | 9056223 | ||
| BC147176.1 | - | 2 | 144 | 9056216 | ||
| DQ316772.1 | - | 2 | 128 | |||

Dmrtc1a
Genomic Map