siRNA Details
| Gene Symbol: Fam71d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027597.4 | NP_081873.2 | 3, 4 | 484 | ||
| NM_029069.1 | NP_083345.1 | 3, 4 | 489 | |||
| GenBank mRNA | AB473726.1 | - | 1, 2 | 164 | ||
| AK014851.1 | - | 3, 4 | 490 | |||
| AK015802.1 | - | 3, 4 | 337 | |||
| BC051149.1 | - | 3, 4 | 344 | 6707726 | ||

Fam71d
Genomic Map