siRNA Details
| Gene Symbol: Spata1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027617.3 | NP_081893.1 | 6, 7 | 771 | ||
| GenBank mRNA | BC139279.1 | - | 5, 6 | 684 | 8862301 | |
| BC139280.1 | - | 5, 6 | 684 | 8862302 | ||
| BC171970.1 | - | 5, 6 | 684 | |||

Spata1
Genomic Map