siRNA Details
| Gene Symbol: Fsip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027759.3 | NP_082035.1 | 2, 3 | 297 | ||
| GenBank mRNA | AK005924.1 | - | 2, 3 | 288 | ||
| AK017026.1 | - | 2, 3 | 286 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fsip1| Gene Symbol: Fsip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027759.3 | NP_082035.1 | 2, 3 | 297 | ||
| GenBank mRNA | AK005924.1 | - | 2, 3 | 288 | ||
| AK017026.1 | - | 2, 3 | 286 | |||