siRNA Details
| Gene Symbol: Stk11ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027886.3 | NP_082162.3 | 4 | 445 | ||
| GenBank mRNA | AK004757.2 | - | 4 | 398 | ||
| AK156122.1 | - | 3 | 533 | |||
| AK165911.1 | - | 4 | 436 | |||
| AK171989.1 | - | 3 | 307 | |||
| AK190662.1 | - | 2 | 239 | |||
| AK210261.1 | - | 2 | 159 | |||
| BC042403.1 | - | 3 | 304 | |||
| BC061635.1 | - | 4 | 428 | 6530178 | ||

Stk11ip
Genomic Map