siRNA Details
| Gene Symbol: Susd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027890.5 | NP_082166.3 | 2 | 181 | ||
| GenBank mRNA | AK004703.2 | - | 2 | 151 | ||
| AK028817.1 | - | 2 | 157 | |||
| AK037059.1 | - | 2 | 163 | |||
| AK037127.1 | - | 2 | 132 | |||
| AK038944.1 | - | 2 | 182 | |||
| AK042469.1 | - | 2 | 165 | |||
| AK128959.1 | - | 2 | 153 | |||
| AK167155.1 | - | 2 | 182 | |||
| BB632717.1 | - | 2 | 182 | |||
| BC026837.1 | - | 2 | 134 | 4222578 | ||

Susd2
Genomic Map