siRNA Details
| Gene Symbol: Selo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027905.2 | NP_082181.2 | 4, 5 | 1089 | ||
| GenBank mRNA | AK005048.1 | - | 4, 5 | 1089 | ||
| AK054432.1 | - | 4, 5 | 1051 | |||
| AK088096.1 | - | 4, 5 | 1056 | |||
| AK161059.1 | - | 4, 5 | 1040 | |||
| BC048849.1 | - | 1, 2 | 112 | |||
| BC117817.1 | - | 5, 6 | 604 | |||
| BC117818.1 | - | 5, 6 | 637 | |||
| BC141293.1 | - | 4, 5 | 1158 | 9055905 | ||

Selo
Genomic Map