siRNA Details
| Gene Symbol: Fam71d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027597.4 | NP_081873.2 | 4 | 497 | ||
| NM_029069.1 | NP_083345.1 | 4 | 502 | |||
| GenBank mRNA | AB473726.1 | - | 2 | 177 | ||
| AK014851.1 | - | 4 | 503 | |||
| AK015802.1 | - | 4 | 350 | |||
| BC051149.1 | - | 4 | 357 | 6707726 | ||

Fam71d
Genomic Map