siRNA Details
Gene Symbol: Tex19.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027622.3 | NP_081898.1 | 3 | 904 | ||
GenBank mRNA | AK014985.1 | - | 3 | 923 | ||
BC141309.1 | - | 3 | 494 | 9055921 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tex19.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027622.3 | NP_081898.1 | 3 | 904 | ||
GenBank mRNA | AK014985.1 | - | 3 | 923 | ||
BC141309.1 | - | 3 | 494 | 9055921 |