siRNA Details
| Gene Symbol: 4931428L18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016212.1 | - | 2 | 309 | ||
| AK016483.1 | - | 2 | 264 | |||
| AK162175.1 | - | 2 | 263 | |||
| BC147514.1 | - | 2 | 280 | 9056506 | ||
| BC147546.1 | - | 2 | 280 | 9056537 | ||
| BC172083.1 | - | 2 | 280 | 9054369 | ||

4931428L18Rik
Genomic Map