siRNA Details
| Gene Symbol: Dhrs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027790.2 | NP_082066.2 | 5 | 453 | ||
| GenBank mRNA | AK017274.1 | - | 5 | 455 | ||
| BC117731.2 | - | 4 | 346 | 40089799 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhrs2| Gene Symbol: Dhrs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027790.2 | NP_082066.2 | 5 | 453 | ||
| GenBank mRNA | AK017274.1 | - | 5 | 455 | ||
| BC117731.2 | - | 4 | 346 | 40089799 | ||