siRNA Details
| Gene Symbol: Rnf215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027859.2 | NP_082135.2 | 2 | 529 | ||
| GenBank mRNA | AK002414.1 | - | 2 | 487 | ||
| AK047194.1 | - | 477 | ||||
| AK154140.1 | - | 2 | 512 | |||
| BC103627.1 | - | 2 | 453 | 6486672 | ||
| BC115907.1 | - | 2 | 476 | 40105152 | ||
| BC115908.1 | - | 2 | 476 | 40105148 | ||

Rnf215
Genomic Map