siRNA Details
| Gene Symbol: Gypc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048207.1 | NP_001041672.1 | 3 | 731 | ||
| GenBank mRNA | AK002769.1 | - | 3 | 561 | ||
| AK082930.1 | - | 3 | 645 | |||
| AK152607.1 | - | 3 | 661 | |||
| AK159730.1 | - | 3 | 665 | |||
| AK162472.1 | - | 2 | 464 | |||
| AY234183.1 | - | 3 | 731 | |||
| BC027408.1 | - | 3 | 671 | 4952917 | ||

Gypc
Genomic Map