siRNA Details
| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF363690.1 | - | 4 | 609 | ||
| AK147276.1 | - | 4 | 1242 | |||
| AK147535.1 | - | 5 | 3360 | |||
| BC058665.1 | - | 1 | 127 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cic| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF363690.1 | - | 4 | 609 | ||
| AK147276.1 | - | 4 | 1242 | |||
| AK147535.1 | - | 5 | 3360 | |||
| BC058665.1 | - | 1 | 127 | |||