siRNA Details
| Gene Symbol: Lrwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027891.4 | NP_082167.2 | 8 | 1122 | ||
| GenBank mRNA | AK004717.1 | - | 6 | 2084 | ||
| AK051910.1 | - | 8 | 1070 | |||
| AK084948.1 | - | 8 | 1122 | |||
| AK153976.1 | - | 8 | 1093 | |||
| AK154352.1 | - | 8 | 1064 | |||
| AK167020.1 | - | 8 | 1071 | |||
| AK172708.1 | - | 8 | 1090 | |||
| BC048397.1 | - | 8 | 1101 | 6396815 | ||

Lrwd1
Genomic Map