siRNA Details
| Gene Symbol: Ints10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293791.1 | NP_001280720.1 | 7 | 771 | ||
| NM_001293792.1 | NP_001280721.1 | 7 | 771 | |||
| NM_027590.4 | NP_081866.1 | 7 | 771 | |||
| GenBank mRNA | AK014939.1 | - | 7 | 747 | ||
| AK030974.1 | - | 7 | 735 | |||
| AK034498.1 | - | 7 | 687 | |||
| AK040574.1 | - | 7 | 772 | |||
| AK047093.1 | - | 7 | 766 | |||
| AK147743.1 | - | 7 | 745 | |||
| BC031923.1 | - | 7 | 734 | 4486323 | ||

Ints10
Genomic Map