siRNA Details
| Gene Symbol: Ccdc178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027616.3 | NP_081892.2 | 5 | 310 | ||
| GenBank mRNA | AK014977.1 | - | 6 | 479 | ||
| AK029534.1 | - | 5 | 311 | |||
| BC140993.1 | - | 5 | 348 | 9055605 | ||
| BC145066.1 | - | 5 | 348 | 9053614 | ||
| BC145069.1 | - | 5 | 355 | 9053617 | ||

Ccdc178
Genomic Map